| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600011385.mm.1 |
NM_001170395.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011385.mm.1 |
NM_001170395.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011385.mm.1 |
NM_053094.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011385.mm.1 |
NM_053094.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011385.mm.1 |
TC0600001418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600011383.mm.1 |
NM_001170395.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011383.mm.1 |
NM_001170395.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011383.mm.1 |
NM_053094.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011383.mm.1 |
NM_053094.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011383.mm.1 |
TC0600001418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600011384.mm.1 |
NM_001170395.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011384.mm.1 |
NM_001170395.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011384.mm.1 |
NM_053094.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011384.mm.1 |
NM_053094.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011384.mm.1 |
TC0600001418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600011386.mm.1 |
NM_001170395.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011386.mm.1 |
NM_001170395.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011386.mm.1 |
NM_053094.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011386.mm.1 |
NM_053094.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600011386.mm.1 |
TC0600001418.mm.1 |
MTA TC |