Assay Details
| Gene Symbol: Rims3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047745.1 | - | 1 - 2 | 309 | 93 | |
| AK136831.1 | - | 1 - 2 | 88 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK136831.1 transcripts.
Set Membership: |
|

Rims3
Genomic Map