Assay Details
| Gene Symbol: Rims3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043152.1 | - | 2 - 3 | 463 | 53 | |
| AK047745.1 | - | 2 - 3 | 479 | 53 | ||
| AK136831.1 | - | 2 - 3 | 258 | 53 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK136831.1 transcripts.
Set Membership: |
|

Rims3
Genomic Map