Assay Details
| Gene Symbol: Sesn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013370.2 | NP_001013388.2 | 5 - 6 | 848 | 98 | |
| NM_001162908.1 | NP_001156380.1 | 5 - 6 | 1418 | 98 | ||
| GenBank mRNA | AK159531.1 | - | 5 - 6 | 794 | 98 | |
| AK160083.1 | - | 5 - 6 | 809 | 98 | ||
| BC055753.1 | - | 5 - 6 | 838 | 6414521 | 98 | |
| BC100403.1 | - | 5 - 6 | 1418 | 30432354 | 98 | |
| KF543242.1 | - | 5 - 6 | 794 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sesn1
Genomic Map