Assay Details
| Gene Symbol: Alpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287172.1 | NP_001274101.1 | 2 - 3 | 329 | 66 | |
| NM_001287176.1 | NP_001274105.1 | 2 - 3 | 452 | 66 | ||
| NM_007431.3 | NP_031457.2 | 2 - 3 | 304 | 66 | ||
| GenBank mRNA | AB473959.1 | - | 1 - 2 | 68 | 66 | |
| AK076459.1 | - | 2 - 3 | 239 | 66 | ||
| AK161780.1 | - | 2 - 3 | 240 | 66 | ||
| AK164002.1 | - | 2 - 3 | 297 | 66 | ||
| AK166732.1 | - | 2 - 3 | 238 | 66 | ||
| AK167023.1 | - | 2 - 3 | 239 | 66 | ||
| AK167481.1 | - | 2 - 3 | 239 | 66 | ||
| BC065175.1 | - | 2 - 3 | 227 | 6807509 | 66 | |
| J02980.1 | - | 2 - 3 | 243 | 66 | ||
| X13409.1 | - | 2 - 3 | 187 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alpl
Genomic Map