Assay Details
Gene Symbol: Alpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287172.1 | NP_001274101.1 | 3 - 4 | 454 | 71 | |
NM_001287176.1 | NP_001274105.1 | 3 - 4 | 577 | 71 | ||
NM_007431.3 | NP_031457.2 | 3 - 4 | 429 | 71 | ||
GenBank mRNA | AB473959.1 | - | 2 - 3 | 193 | 71 | |
AK076459.1 | - | 3 - 4 | 364 | 71 | ||
AK161780.1 | - | 3 - 4 | 365 | 71 | ||
AK164002.1 | - | 3 - 4 | 422 | 71 | ||
AK166732.1 | - | 3 - 4 | 363 | 71 | ||
AK167023.1 | - | 3 - 4 | 364 | 71 | ||
AK167481.1 | - | 3 - 4 | 364 | 71 | ||
BC065175.1 | - | 3 - 4 | 352 | 6807509 | 71 | |
CN539670.1 | - | 3 - 4 | 456 | 71 | ||
J02980.1 | - | 3 - 4 | 368 | 71 | ||
X13409.1 | - | 3 - 4 | 312 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
