| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100000141.mm.1 |
NM_001253690.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000141.mm.1 |
NM_001253690.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000141.mm.1 |
NM_001253691.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000141.mm.1 |
NM_001253691.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000141.mm.1 |
NM_019574.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000141.mm.1 |
NM_019574.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000141.mm.1 |
NR_045108.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000141.mm.1 |
NR_045108.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000141.mm.1 |
NR_045109.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000141.mm.1 |
NR_045109.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000141.mm.1 |
TC1100000007.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000143.mm.1 |
NM_001253690.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000143.mm.1 |
NM_001253690.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000143.mm.1 |
NR_045108.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000143.mm.1 |
NR_045108.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000143.mm.1 |
TC1100000007.mm.1 |
MTA TC |