Assay Details
| Gene Symbol: Bsdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133889.2 | NP_598650.2 | 5 - 6 | 423 | 60 | |
| GenBank mRNA | AK037201.1 | - | 5 - 6 | 419 | 60 | |
| AK144053.1 | - | 5 - 6 | 418 | 60 | ||
| BC021480.1 | - | 4 - 5 | 403 | 60 | ||
| BC049111.1 | - | 5 - 6 | 423 | 6415574 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bsdc1
Genomic Map