Assay Details
| Gene Symbol: Bsdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133889.2 | NP_598650.2 | 8 - 9 | 690 | 85 | |
| GenBank mRNA | AK037201.1 | - | 8 - 9 | 686 | 85 | |
| AK144053.1 | - | 8 - 9 | 685 | 85 | ||
| BC021480.1 | - | 7 - 8 | 670 | 85 | ||
| BC049111.1 | - | 8 - 9 | 690 | 6415574 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bsdc1
Genomic Map