Assay Details
| Gene Symbol: Tctex1d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175030.2 | NP_778195.1 | 3 - 4 | 219 | 60 | |
| GenBank mRNA | AK029345.1 | - | 3 - 4 | 220 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029345.1, NM_175030.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097372 | NM_175030.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097372 | AK029345.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097372 | BC115474.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324559 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509138 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133555 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097372 | NM_175030.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097372 | AK029345.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097372 | BC115474.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23232603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23232604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23232602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23232605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23232600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23232606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23232597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23232599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23232596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23232607 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133555 | - | - |
| Whole Mouse Genome G4122A | A_52_P640611 | NM_175030.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P640611 | AK029345.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122797 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133555 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03611404 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03611412 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03611383 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03611392 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03611397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23232603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03376853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03611399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03709602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03871049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03871047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03601771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03611415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03871046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03376856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03245457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03611382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03732270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03216951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03732268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03898363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03216953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232568 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03871044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03245456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23232552 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10507459 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507460 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507458 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507457 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436653_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10507456 | - | - |
| Mouse Genome 430 2.0 Array | 1436653_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436653_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 242646_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 242646_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400009618.mm.1 | NM_175030.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400009618.mm.1 | NM_175030.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400009618.mm.1 | TC0400001278.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436653_PM_at | - | - |
| Mouse Expression Set 430 | 1436653_at | - | - |
| Murine Genome U74v2 Set | 166617_at | - | - |
| Murine Genome U74v2 Set | 166617_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436653_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436653_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436653_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436653_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 577596 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1410450 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1410450 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001410450 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214031 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214031 | - | - |
| mouse-6 v1.1 (Illumina) | scl25056.4.1_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1410450 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25056.4.1_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1410450 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1214031 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214031 | - | - |

Tctex1d4
Genomic Map