Assay Details
| Gene Symbol: Hbs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042593.1 | NP_001036058.1 | 10 - 11 | 1442 | 61 | |
| NM_019702.2 | NP_062676.2 | 10 - 11 | 1451 | 61 | ||
| GenBank mRNA | AF087672.1 | - | 7 - 8 | 1312 | 61 | |
| AK146678.1 | - | 10 - 11 | 1408 | 61 | ||
| AK166140.1 | - | 10 - 11 | 1405 | 61 | ||
| AK168671.1 | - | 10 - 11 | 1405 | 61 | ||
| AK169281.1 | - | 10 - 11 | 1405 | 61 | ||
| AK173091.1 | - | 10 - 11 | 1397 | 61 | ||
| BC010251.1 | - | 10 - 11 | 1376 | 3964560 | 61 | |
More Information
Set Membership: |
|

Hbs1l
Genomic Map