Assay Details
| Gene Symbol: Hbs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042593.1 | NP_001036058.1 | 12 - 13 | 1625 | 78 | |
| NM_019702.2 | NP_062676.2 | 12 - 13 | 1634 | 78 | ||
| GenBank mRNA | AF087672.1 | - | 9 - 10 | 1495 | 78 | |
| AK146678.1 | - | 12 - 13 | 1591 | 78 | ||
| AK166140.1 | - | 12 - 13 | 1588 | 78 | ||
| AK168671.1 | - | 12 - 13 | 1588 | 78 | ||
| AK169281.1 | - | 12 - 13 | 1588 | 78 | ||
| AK173091.1 | - | 12 - 13 | 1580 | 78 | ||
| BC010251.1 | - | 12 - 13 | 1559 | 3964560 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hbs1l
Genomic Map