Assay Details
| Gene Symbol: Smpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009213.2 | NP_033239.1 | 9 - 10 | 1187 | 89 | |
| GenBank mRNA | AJ222800.1 | - | 9 - 10 | 1188 | 89 | |
| AK075616.1 | - | 9 - 10 | 1197 | 89 | ||
| AK088962.1 | - | 6 - 7 | 1955 | 89 | ||
| AK161573.1 | - | 2299 | 89 | |||
| BC010978.1 | - | 9 - 10 | 1197 | 4193282 | 89 | |
| BC086487.1 | - | 2 - 3 | 2333 | 89 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089555 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089558 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089554 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089557 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089553 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137654 | BC086487.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137654 | AK075616.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137654 | AK088962.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137654 | AK161573.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137654 | BC010978.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313674 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46025 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106036 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137654 | BC086487.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137654 | AK075616.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137654 | AK088962.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137654 | AK161573.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137654 | BC010978.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234421 | AK075616.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234421 | AK088962.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234421 | BC010978.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234421 | AJ222800.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234421 | NM_009213.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089548 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106036 | - | - |
| Whole Mouse Genome G4122A | A_51_P234421 | AK075616.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P234421 | AK088962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P234421 | BC010978.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P234421 | AJ222800.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P234421 | NM_009213.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137654 | BC086487.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137654 | AK075616.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137654 | AK088962.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137654 | AK161573.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137654 | BC010978.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135424 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106036 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08523378 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08523385 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08234988 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08523387 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08234990 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089553 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089554 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089555 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089558 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08838616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08838608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08523375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08838619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089551 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10368812 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368814 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368809 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368813 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368808 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368815 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368810 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368807 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368816 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368811 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416999_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10368806 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10362794 | - | - |
| Mouse Genome 430 2.0 Array | 1416999_at | - | - |
| Mouse Genome 430 2.0 Array | 1416999_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20598_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20598_PM_at | - | - |
| Mouse Expression Set 430 | 1416999_PM_at | - | - |
| Mouse Expression Set 430 | 1416999_at | - | - |
| Murine Genome U74v2 Set | 101940_PM_at | - | - |
| Murine Genome U74v2 Set | 101940_at | - | - |
| Murine Genome U74v2 Set | 168953_PM_f_at | - | - |
| Murine Genome U74v2 Set | 168953_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416999_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416999_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416999_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416999_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 441733 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2370133 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002370133 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2730161 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2730161 | - | - |
| mouse-6 v1.1 (Illumina) | scl0020598.1_244-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2370133 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0020598.1_244-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2370133 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2867033 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2867033 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0020598.1_244-S | - | - |

Smpd2
Genomic Map