Assay Details
| Gene Symbol: Stk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009288.2 | NP_033314.2 | 11 - 12 | 1941 | 76 | |
| GenBank mRNA | AK148118.1 | - | 11 - 12 | 1884 | 76 | |
| AK220448.1 | - | 11 - 12 | 1863 | 76 | ||
| BC069840.1 | - | 11 - 12 | 2014 | 76 | ||
| BC096624.1 | - | 11 - 12 | 2026 | 76 | ||
| BC128363.1 | - | 11 - 12 | 1827 | 40111177 | 76 | |
| D89728.1 | - | 11 - 12 | 1902 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk10
Genomic Map