Assay Details
| Gene Symbol: Stk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009288.2 | NP_033314.2 | 15 - 16 | 2470 | 56 | |
| GenBank mRNA | AK220448.1 | - | 15 - 16 | 2392 | 56 | |
| BC069840.1 | - | 15 - 16 | 2543 | 56 | ||
| BC096624.1 | - | 15 - 16 | 2555 | 56 | ||
| BC128363.1 | - | 15 - 16 | 2356 | 40111177 | 56 | |
| D89728.1 | - | 15 - 16 | 2431 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk10
Genomic Map