Assay Details
| Gene Symbol: Stk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009288.2 | NP_033314.2 | 17 - 18 | 2789 | 66 | |
| GenBank mRNA | AK220448.1 | - | 17 - 18 | 2711 | 66 | |
| BC069840.1 | - | 17 - 18 | 2862 | 66 | ||
| BC096624.1 | - | 17 - 18 | 2874 | 66 | ||
| BC128363.1 | - | 17 - 18 | 2675 | 40111177 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stk10
Genomic Map