| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000013167.mm.1 |
NM_001033258.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000013167.mm.1 |
NM_001033258.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000013167.mm.1 |
TC1000001809.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000013168.mm.1 |
NM_001033258.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000013168.mm.1 |
NM_001033258.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000013168.mm.1 |
TC1000001809.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000013166.mm.1 |
NM_001033258.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000013166.mm.1 |
NM_001033258.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000013166.mm.1 |
TC1000001809.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000013169.mm.1 |
NM_001033258.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000013169.mm.1 |
NM_001033258.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000013169.mm.1 |
TC1000001809.mm.1 |
MTA TC |