Assay Details
| Gene Symbol: Ago3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153402.2 | NP_700451.2 | 7 - 8 | 1133 | 99 | |
| GenBank mRNA | AB081473.1 | - | 7 - 8 | 985 | 99 | |
| AK035042.1 | - | 7 - 8 | 1099 | 99 | ||
| AK083652.1 | - | 7 - 8 | 1100 | 99 | ||
| AK154436.1 | - | 7 - 8 | 1134 | 99 | ||
| AK171118.1 | - | 7 - 8 | 1134 | 99 | ||
| BC137964.1 | - | 7 - 8 | 980 | 8860983 | 99 | |
| BC145061.1 | - | 7 - 8 | 980 | 99 | ||
| BC145063.1 | - | 6 - 7 | 843 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ago3
Genomic Map