Assay Details
| Gene Symbol: Ago3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153402.2 | NP_700451.2 | 9 - 10 | 1409 | 94 | |
| GenBank mRNA | AK035042.1 | - | 9 - 10 | 1375 | 94 | |
| AK083652.1 | - | 9 - 10 | 1376 | 94 | ||
| AK154436.1 | - | 9 - 10 | 1410 | 94 | ||
| AK171118.1 | - | 9 - 10 | 1410 | 94 | ||
| BC137964.1 | - | 9 - 10 | 1256 | 8860983 | 94 | |
| BC145061.1 | - | 9 - 10 | 1256 | 94 | ||
| BC145063.1 | - | 8 - 9 | 1119 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ago3
Genomic Map