Assay Details
| Gene Symbol: Tmem53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285814.1 | NP_001272743.1 | 2 - 3 | 265 | 83 | |
| NM_029589.1 | NP_083865.1 | 2 - 3 | 101 | 83 | ||
| GenBank mRNA | AK005885.1 | - | 2 - 3 | 105 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005885.1, NM_001285814.1, NM_029589.1 transcripts.
Set Membership: |
|

Tmem53
Genomic Map