Assay Details
| Gene Symbol: Tctex1d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC115474.2 | - | 2 - 3 | 82 | 40100473 | 57 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC115474.2 transcripts.
Set Membership: |
|

Tctex1d4
Genomic Map