Assay Details
| Gene Symbol: Tctex1d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175030.2 | NP_778195.1 | 2 - 3 | 152 | 83 | |
| GenBank mRNA | AK029345.1 | - | 2 - 3 | 153 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029345.1, NM_175030.2 transcripts.
Set Membership: |
|

Tctex1d4
Genomic Map