Assay Details
| Gene Symbol: Rwdd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025614.3 | NP_079890.1 | 1 - 2 | 250 | 86 | |
| GenBank mRNA | AF503942.1 | - | 1 - 2 | 142 | 86 | |
| AK005278.1 | - | 1 - 2 | 162 | 86 | ||
| AK009510.1 | - | 1 - 2 | 119 | 86 | ||
| AK011277.1 | - | 1 - 2 | 250 | 86 | ||
| AK012504.1 | - | 1 - 2 | 221 | 86 | ||
| BC062136.1 | - | 1 - 2 | 132 | 6508909 | 86 | |
More Information
Set Membership: |
|

Rwdd1
Genomic Map