Assay Details
| Gene Symbol: Ccdc162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK137750.1 | - | 5 - 6 | 781 | 67 | |
| BC112401.1 | - | 12 - 13 | 1666 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc162| Gene Symbol: Ccdc162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK137750.1 | - | 5 - 6 | 781 | 67 | |
| BC112401.1 | - | 12 - 13 | 1666 | 67 | ||
Set Membership: |
|