Assay Details
| Gene Symbol: Ccdc162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050292.1 | - | 5 - 6 | 622 | 66 | |
| BC112401.1 | - | 16 - 17 | 2239 | 66 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177248 | AK137750.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114367 | BC112401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114367 | AK050292.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114367 | AK137750.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114367 | NM_001177571.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172851 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163517 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177248 | AK137750.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114367 | BC112401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114367 | AK050292.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114367 | AK137750.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114367 | NM_001177571.1 | mouse_transcriptome |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08540708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08685266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08933602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08662429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08685273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08668089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08668091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08439601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08636999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08895910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08685275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08888102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08507295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08439602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08259356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08924995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08452231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08402115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08408296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08380205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08950871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08685274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08685270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08259344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08694896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08288799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08939798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08284191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08259343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27090111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089948 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10368819 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368834 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368823 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368831 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368820 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368827 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368835 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368832 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368829 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368824 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368818 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368822 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368830 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368826 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368825 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368833 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445787_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10368817 | - | - |
| Mouse Genome 430 2.0 Array | 1445787_at | - | - |
| Mouse Genome 430 2.0 Array | 1445787_PM_at | - | - |
| Mouse Expression Set 430 | 1445787_PM_at | - | - |
| Mouse Expression Set 430 | 1445787_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445787_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445787_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236488 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2948241 | - | - |

Ccdc162
Genomic Map