| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000014548.mm.1 |
NM_027879.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014548.mm.1 |
NM_027879.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014548.mm.1 |
TC1000002046.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000014547.mm.1 |
NM_027879.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014547.mm.1 |
NM_027879.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014547.mm.1 |
TC1000002046.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000014544.mm.1 |
NM_027879.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014544.mm.1 |
NM_027879.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014544.mm.1 |
TC1000002046.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000014546.mm.1 |
NM_027879.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014546.mm.1 |
NM_027879.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014546.mm.1 |
TC1000002046.mm.1 |
MTA TC |