Assay Details
| Gene Symbol: Serbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113565.1 | NP_001107037.1 | 4 - 5 | 812 | 81 | |
| NM_001113566.1 | NP_001107038.1 | 4 - 5 | 794 | 81 | ||
| GenBank mRNA | AK010860.1 | - | 4 - 5 | 812 | 81 | |
| AK088278.1 | - | 4 - 5 | 763 | 81 | ||
| AK131990.1 | - | 4 - 5 | 763 | 81 | ||
| AK135473.1 | - | 3 - 4 | 594 | 81 | ||
| AK139670.1 | - | 4 - 5 | 781 | 81 | ||
| AK149505.1 | - | 4 - 5 | 766 | 81 | ||
| BC006030.1 | - | 4 - 5 | 766 | 81 | ||
| BC013665.1 | - | 4 - 5 | 748 | 4193513 | 81 | |
| BC030502.1 | - | 4 - 5 | 756 | 5401573 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Serbp1
Genomic Map