Assay Details
| Gene Symbol: Chmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025783.3 | NP_080059.2 | 2 - 3 | 235 | 113 | |
| GenBank mRNA | AK009414.1 | - | 2 - 3 | 109 | 113 | |
| AK014818.1 | - | 2 - 3 | 173 | 113 | ||
| AK016677.1 | - | 2 - 3 | 159 | 113 | ||
| AK018611.1 | - | 2 - 3 | 190 | 113 | ||
| AK083562.1 | - | 2 - 3 | 236 | 113 | ||
| BC027441.1 | - | 2 - 3 | 99 | 113 | ||
| BC039759.1 | - | 2 - 3 | 110 | 113 | ||
| BC049964.1 | - | 2 - 3 | 156 | 3154282 | 113 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Chmp3
Genomic Map