Assay Details
| Gene Symbol: Chmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025783.3 | NP_080059.2 | 1 - 2 | 175 | 105 | |
| GenBank mRNA | AK009414.1 | - | 1 - 2 | 49 | 105 | |
| AK014818.1 | - | 1 - 2 | 113 | 105 | ||
| AK016677.1 | - | 1 - 2 | 99 | 105 | ||
| AK018611.1 | - | 1 - 2 | 130 | 105 | ||
| AK083562.1 | - | 1 - 2 | 176 | 105 | ||
| BC049964.1 | - | 1 - 2 | 96 | 3154282 | 105 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK009414.1,AK014818.1,AK016677.1,AK018611.1,AK083562.1,BC049964.1,NM_025783.3 transcripts.
This assay is in the 5'UTR region of the AK009414.1,AK014818.1,AK016677.1,AK018611.1,AK083562.1,BC049964.1,NM_025783.3 transcripts.
Set Membership: |
|

Chmp3
Genomic Map