Assay Details
| Gene Symbol: Chmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025783.3 | NP_080059.2 | 4 - 5 | 537 | 105 | |
| GenBank mRNA | AK009414.1 | - | 4 - 5 | 411 | 105 | |
| AK014818.1 | - | 4 - 5 | 475 | 105 | ||
| AK016677.1 | - | 4 - 5 | 461 | 105 | ||
| AK083562.1 | - | 4 - 5 | 538 | 105 | ||
| BC027441.1 | - | 4 - 5 | 401 | 105 | ||
| BC039759.1 | - | 4 - 5 | 412 | 105 | ||
| BC049964.1 | - | 4 - 5 | 458 | 3154282 | 105 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Chmp3
Genomic Map