Assay Details
| Gene Symbol: Gm5420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045843.1 | 2 - 3 | 627 | 60 | ||
| GenBank mRNA | AK090213.1 | - | 2 - 3 | 627 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P779862 | AK090213.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P779862 | AK090213.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P779862 | AK090213.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P779862 | AK090213.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P779862 | AK090213.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P779862 | AK090213.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109472 | - | - |
| Agilent Mouse cDNA Microarray (G4104A) | 1025464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08798063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08741073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08729680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08925122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08741075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08523034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08633680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08523024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08553886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08925121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08523023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08905999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08523033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08347356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08523026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08843302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08523020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08729686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08905998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08741056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08523039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26994622 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR1000001228.mm.1 | NR_045843.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001228.mm.1 | NR_045843.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001228.mm.1 | TC1000000170.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 300771 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104590348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104590348 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1256940 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104590348 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104590348 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2957044 | - | - |

Gm5420
Genomic Map