Assay Details
| Gene Symbol: Cyp4x1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK048538.1 | - | 1 - 2 | 577 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
cytochrome P450, family 4, subfamily x, polypeptide 1 |
Cyp4x1 |
A230025G20, CYPIVX1, CYP_a, Cyp4a28-ps |
Anti-sense | Chr. 4 - 115108682 - 115133977 on NCBI Build GRCm38 | |
| Downstream |
cytochrome P450, family 4, subfamily a, polypeptide 29 |
Cyp4a29 |
Cyp4a29-ps, OTTMUSG00000008612 |
107.5kb | Sense | Chr. 4 - 115242084 - 115254557 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Cyp4x1os
Genomic Map