| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1453308_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10373751 |
- |
- |
| Mouse Genome 430 2.0 Array |
1453308_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1453308_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1100000207.mm.1 |
NR_028317.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000207.mm.1 |
NR_028317.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000207.mm.1 |
NR_045275.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000207.mm.1 |
NR_045275.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000207.mm.1 |
NR_045276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000207.mm.1 |
NR_045276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000207.mm.1 |
TC1100000011.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000213.mm.1 |
NR_045275.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000213.mm.1 |
NR_045275.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000213.mm.1 |
NR_045276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000213.mm.1 |
NR_045276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000213.mm.1 |
TC1100000011.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000211.mm.1 |
NR_045275.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000211.mm.1 |
NR_045275.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000211.mm.1 |
NR_045276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000211.mm.1 |
NR_045276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000211.mm.1 |
TC1100000011.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000208.mm.1 |
NR_028317.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000208.mm.1 |
NR_028317.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000208.mm.1 |
NR_045276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000208.mm.1 |
NR_045276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000208.mm.1 |
TC1100000011.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000204.mm.1 |
NR_028317.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000204.mm.1 |
NR_028317.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000204.mm.1 |
TC1100000011.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000212.mm.1 |
NR_045275.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000212.mm.1 |
NR_045275.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000212.mm.1 |
NR_045276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000212.mm.1 |
NR_045276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000212.mm.1 |
TC1100000011.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000214.mm.1 |
NR_045275.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000214.mm.1 |
NR_045275.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000214.mm.1 |
NR_045276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000214.mm.1 |
NR_045276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000214.mm.1 |
TC1100000011.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000219.mm.1 |
NR_028317.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000219.mm.1 |
NR_028317.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000219.mm.1 |
TC1100000011.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000209.mm.1 |
NR_028317.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000209.mm.1 |
NR_028317.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000209.mm.1 |
NR_045276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000209.mm.1 |
NR_045276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000209.mm.1 |
TC1100000011.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000218.mm.1 |
NR_028317.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000218.mm.1 |
NR_028317.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000218.mm.1 |
TC1100000011.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000220.mm.1 |
NR_028317.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000220.mm.1 |
NR_028317.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000220.mm.1 |
TC1100000011.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1453308_PM_at |
- |
- |
| Mouse Expression Set 430 |
1453308_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453308_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453308_PM_at |
- |
- |