Assay Details
| Gene Symbol: Bclaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025392.1 | NP_001020563.1 | 7 - 8 | 2201 | 66 | |
| NM_001025393.1 | NP_001020564.1 | 7 - 8 | 2201 | 66 | ||
| NM_153787.2 | NP_722482.1 | 7 - 8 | 2195 | 66 | ||
| GenBank mRNA | AK079110.1 | - | 3 - 4 | 451 | 66 | |
| AK088741.1 | - | 7 - 8 | 2136 | 66 | ||
| AK129071.1 | - | 4 - 5 | 1485 | 66 | ||
| AK141834.1 | - | 7 - 8 | 2148 | 66 | ||
| AK149724.1 | - | 7 - 8 | 2154 | 66 | ||
| AK150017.1 | - | 7 - 8 | 2154 | 66 | ||
| AK161608.1 | - | 1090 | 66 | |||
| AK161692.1 | - | 7 - 8 | 2199 | 66 | ||
| BC029682.1 | - | 7 - 8 | 2132 | 66 | ||
| BC034300.1 | - | 7 - 8 | 2127 | 4507876 | 66 | |
| BC086624.1 | - | 7 - 8 | 2183 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bclaf1
Genomic Map