| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100000287.mm.1 |
NM_001159288.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000287.mm.1 |
NM_001159288.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000287.mm.1 |
NM_198162.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000287.mm.1 |
NM_198162.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000287.mm.1 |
TC1100000021.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000283.mm.1 |
NM_001159288.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000283.mm.1 |
NM_001159288.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000283.mm.1 |
NM_198162.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000283.mm.1 |
NM_198162.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000283.mm.1 |
TC1100000021.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000285.mm.1 |
NM_198162.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000285.mm.1 |
NM_198162.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000285.mm.1 |
TC1100000021.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000286.mm.1 |
NM_001159288.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000286.mm.1 |
NM_001159288.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000286.mm.1 |
NM_198162.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000286.mm.1 |
NM_198162.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000286.mm.1 |
TC1100000021.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000278.mm.1 |
NM_001159288.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000278.mm.1 |
NM_001159288.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000278.mm.1 |
TC1100000021.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000279.mm.1 |
NM_001159288.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000279.mm.1 |
NM_001159288.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000279.mm.1 |
TC1100000021.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000281.mm.1 |
NM_001159288.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000281.mm.1 |
NM_001159288.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000281.mm.1 |
TC1100000021.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000282.mm.1 |
NM_001159288.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000282.mm.1 |
NM_001159288.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000282.mm.1 |
TC1100000021.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000284.mm.1 |
NM_001159288.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000284.mm.1 |
NM_001159288.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000284.mm.1 |
NM_198162.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000284.mm.1 |
NM_198162.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000284.mm.1 |
TC1100000021.mm.1 |
MTA TC |