| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100000294.mm.1 |
NM_001159288.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000294.mm.1 |
NM_001159288.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000294.mm.1 |
NM_198162.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000294.mm.1 |
NM_198162.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000294.mm.1 |
TC1100000021.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000296.mm.1 |
NM_001159288.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000296.mm.1 |
NM_001159288.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000296.mm.1 |
NM_198162.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000296.mm.1 |
NM_198162.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000296.mm.1 |
TC1100000021.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100000295.mm.1 |
NM_001159288.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000295.mm.1 |
NM_001159288.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000295.mm.1 |
NM_198162.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000295.mm.1 |
NM_198162.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100000295.mm.1 |
TC1100000021.mm.1 |
MTA TC |