Assay Details
| Gene Symbol: Lage3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025410.2 | NP_079686.1 | 2 - 3 | 418 | 76 | |
| GenBank mRNA | AK002539.1 | - | 2 - 3 | 419 | 76 | |
| AK004210.1 | - | 2 - 3 | 411 | 76 | ||
| BC027305.1 | - | 2 - 3 | 384 | 4009230 | 76 | |
More Information
Set Membership: |
|

Lage3
Genomic Map