Assay Details
| Gene Symbol: Fignl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163359.1 | NP_001156831.1 | 1 - 2 | 84 | 63 | |
| GenBank mRNA | AK143850.1 | - | 1 - 2 | 53 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK143850.1, NM_001163359.1 transcripts.
Set Membership: |
|

Fignl1
Genomic Map