Assay Details
| Gene Symbol: Hmgcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008254.2 | NP_032280.2 | 1 - 2 | 91 | 75 | |
| GenBank mRNA | AK077842.1 | - | 1 - 2 | 80 | 75 | |
| AK132035.1 | - | 1 - 2 | 80 | 75 | ||
| AK144768.1 | - | 1 - 2 | 89 | 75 | ||
| AK145251.1 | - | 1 - 2 | 87 | 75 | ||
| AK154033.1 | - | 1 - 2 | 80 | 75 | ||
| BB852743.1 | - | 1 - 2 | 91 | 75 | ||
| BC025440.1 | - | 1 - 2 | 78 | 5049759 | 75 | |
| S65036.1 | - | 1 - 2 | 79 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hmgcl
Genomic Map