Assay Details
| Gene Symbol: Erlec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025745.3 | NP_080021.3 | 4 - 5 | 478 | 100 | |
| GenBank mRNA | AK002471.1 | - | 4 - 5 | 469 | 100 | |
| AK077120.1 | - | 4 - 5 | 471 | 100 | ||
| AK133874.1 | - | 4 - 5 | 479 | 100 | ||
| BC018468.1 | - | 4 - 5 | 488 | 3591901 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Erlec1
Genomic Map