Assay Details
| Gene Symbol: Lmtk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005511.3 | NP_001005511.2 | 8 - 9 | 866 | 68 | |
| NM_001290990.1 | NP_001277919.1 | 7 - 8 | 1021 | 68 | ||
| NR_110987.1 | 8 - 9 | 866 | 68 | |||
| GenBank mRNA | AB288873.1 | - | 8 - 9 | 860 | 68 | |
| BC057032.1 | - | 7 - 8 | 754 | 68 | ||
| BC059845.1 | - | 8 - 9 | 1075 | 68 | ||
| BC083185.1 | - | 7 - 8 | 1021 | 6855048 | 68 | |
| BC094377.1 | - | 8 - 9 | 866 | 30536822 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmtk3
Genomic Map