Assay Details
| Gene Symbol: Sgsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162965.1 | NP_001156437.1 | 16 - 17 | 1867 | 63 | |
| NM_001254731.1 | NP_001241660.1 | 16 - 17 | 1867 | 63 | ||
| NM_172718.3 | NP_766306.2 | 16 - 17 | 1867 | 63 | ||
| NR_027934.1 | 7 - 8 | 1054 | 63 | |||
| NR_027936.1 | 7 - 8 | 1054 | 63 | |||
| GenBank mRNA | AK044716.1 | - | 16 - 17 | 1821 | 63 | |
| AK051018.1 | - | 7 - 8 | 1055 | 63 | ||
| AK053555.1 | - | 16 - 17 | 1801 | 63 | ||
| AK167632.1 | - | 7 - 8 | 1018 | 63 | ||
| AK170021.1 | - | 16 - 17 | 1859 | 63 | ||
| BC138049.1 | - | 16 - 17 | 1793 | 8861068 | 63 | |
| BC138050.1 | - | 16 - 17 | 1793 | 8861069 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgsm1
Genomic Map