| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900014190.mm.1 |
NM_001113408.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014190.mm.1 |
NM_001113408.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014190.mm.1 |
TC1900001594.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900014191.mm.1 |
NM_001113408.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014191.mm.1 |
NM_001113408.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014191.mm.1 |
TC1900001594.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900014185.mm.1 |
NM_001113408.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014185.mm.1 |
NM_001113408.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014185.mm.1 |
NM_010697.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014185.mm.1 |
NM_010697.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014185.mm.1 |
TC1900001594.mm.1 |
MTA TC |