Assay Details
| Gene Symbol: Psd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028627.2 | NP_082903.2 | 10 - 11 | 2216 | 82 | |
| GenBank mRNA | AK220438.1 | - | 5 - 6 | 2333 | 82 | |
| BC043135.1 | - | 6 - 7 | 1045 | 82 | ||
| BC057353.1 | - | 10 - 11 | 2217 | 82 | ||
| BC138652.1 | - | 8 - 9 | 2075 | 8861672 | 82 | |
| BC145352.1 | - | 8 - 9 | 2078 | 9053908 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psd
Genomic Map