Assay Details
| Gene Symbol: Hpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008277.2 | NP_032303.1 | 2 - 3 | 103 | 72 | |
| GenBank mRNA | AK149416.1 | - | 3 - 4 | 141 | 72 | |
| BC013343.1 | - | 2 - 3 | 127 | 4193670 | 72 | |
| D29987.1 | - | 3 - 4 | 141 | 72 | ||
| X59530.1 | - | 2 - 3 | 93 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hpd
Genomic Map