Assay Details
| Gene Symbol: Hpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008277.2 | NP_032303.1 | 3 - 4 | 210 | 58 | |
| GenBank mRNA | AK149416.1 | - | 4 - 5 | 248 | 58 | |
| BC013343.1 | - | 3 - 4 | 234 | 4193670 | 58 | |
| X59530.1 | - | 3 - 4 | 200 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hpd
Genomic Map