Assay Details
| Gene Symbol: Hpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008277.2 | NP_032303.1 | 5 - 6 | 334 | 60 | |
| GenBank mRNA | AK149416.1 | - | 6 - 7 | 372 | 60 | |
| BC013343.1 | - | 5 - 6 | 358 | 4193670 | 60 | |
| D29987.1 | - | 6 - 7 | 372 | 60 | ||
| X59530.1 | - | 5 - 6 | 324 | 60 | ||
More Information
Set Membership: |
|

Hpd
Genomic Map