Assay Details
| Gene Symbol: Hpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008277.2 | NP_032303.1 | 8 - 9 | 601 | 63 | |
| GenBank mRNA | BC013343.1 | - | 8 - 9 | 625 | 4193670 | 63 |
| D29987.1 | - | 9 - 10 | 639 | 63 | ||
| X59530.1 | - | 8 - 9 | 591 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hpd
Genomic Map