Assay Details
| Gene Symbol: Morn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029112.1 | NP_083388.1 | 1 - 2 | 186 | 73 | |
| GenBank mRNA | AK015339.1 | - | 1 - 2 | 339 | 73 | |
| AK077141.1 | - | 1 - 2 | 383 | 73 | ||
| BC019987.1 | - | 1 - 2 | 186 | 4458628 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Morn3
Genomic Map