Assay Details
| Gene Symbol: Morn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029112.1 | NP_083388.1 | 4 - 5 | 693 | 63 | |
| GenBank mRNA | BC019987.1 | - | 4 - 5 | 693 | 4458628 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154809 | NM_029112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154809 | AK015339.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154809 | BC019987.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021423 | NM_029112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021423 | AK015339.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021423 | BC019987.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46576 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821282 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397907 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625654 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724035 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 32967 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145302 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175442 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154809 | NM_029112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154809 | AK015339.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154809 | BC019987.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021423 | NM_029112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021423 | AK015339.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021423 | BC019987.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175442 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01208497 | - | - |
| Whole Mouse Genome G4122A | A_51_P273823 | NM_029112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273823 | BC019987.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P593947 | NM_029112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P593947 | AK015339.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P593947 | AK077141.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P593947 | BC019987.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175442 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04451151 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04451140 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04451148 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04451143 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04451155 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04451156 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04451146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24053636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04439136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04576984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04875745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04799079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04689968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04890515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04844146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04326002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04844149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04451151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04844147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04689973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04451137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04689972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04844148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04367981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04529930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04354900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24053644 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10533602 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533597 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533599 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533601 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533595 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533598 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533596 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430970_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10533594 | - | - |
| Mouse Genome 430 2.0 Array | 1430970_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1430970_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74890_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74890_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500028908.mm.1 | NM_029112.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028908.mm.1 | NM_029112.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028908.mm.1 | TC0500003223.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028902.mm.1 | NM_029112.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028902.mm.1 | NM_029112.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028902.mm.1 | TC0500003223.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028903.mm.1 | NM_029112.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028903.mm.1 | NM_029112.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028903.mm.1 | TC0500003223.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028906.mm.1 | NM_029112.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028906.mm.1 | NM_029112.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028906.mm.1 | TC0500003223.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028905.mm.1 | NM_029112.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028905.mm.1 | NM_029112.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028905.mm.1 | TC0500003223.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430970_PM_a_at | - | - |
| Mouse Expression Set 430 | 1430970_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430970_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430970_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430970_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430970_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 577615 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104010333 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104010333 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223151 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230112 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250365 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104010333 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104010333 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3007767 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3007767 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230112 | - | - |

Morn3
Genomic Map